Assay Details
| Gene Symbol: Ndufs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026688.2 | NP_080964.1 | 4 - 5 | 457 | 66 | |
| GenBank mRNA | AK002501.1 | - | 4 - 5 | 430 | 66 | |
| AK088337.1 | - | 4 - 5 | 458 | 66 | ||
| AK160888.1 | - | 2 - 3 | 2220 | 66 | ||
| BC027270.1 | - | 4 - 5 | 411 | 3491580 | 66 | |
| BC119267.1 | - | 4 - 5 | 411 | 8734016 | 66 | |
| BC119269.1 | - | 4 - 5 | 412 | 8734018 | 66 | |
More Information
Set Membership: |
|

Ndufs3
Genomic Map