Assay Details
| Gene Symbol: Mphosph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026758.3 | NP_081034.1 | 2 - 3 | 187 | 100 | |
| GenBank mRNA | AK003245.1 | - | 2 - 3 | 188 | 100 | |
| BC008161.1 | - | 2 - 3 | 183 | 3964232 | 100 | |
More Information
Set Membership: |
|

Mphosph6
Genomic Map