Assay Details
| Gene Symbol: Ebna1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026932.4 | NP_081208.1 | 4 - 5 | 578 | 86 | |
| GenBank mRNA | AK007491.1 | - | 4 - 5 | 548 | 86 | |
| AK134732.1 | - | 4 - 5 | 533 | 86 | ||
| AK167070.1 | - | 4 - 5 | 533 | 86 | ||
| AK168269.1 | - | 4 - 5 | 505 | 86 | ||
| BC054723.1 | - | 4 - 5 | 561 | 5695176 | 86 | |
| BC062876.1 | - | 4 - 5 | 510 | 5686021 | 86 | |
More Information
Set Membership: |
|

Ebna1bp2
Genomic Map