Assay Details
| Gene Symbol: Ankrd33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164441.1 | NP_001157913.1 | 3 - 4 | 997 | 82 | |
| NM_027496.3 | NP_081772.1 | 3 - 4 | 970 | 82 | ||
| GenBank mRNA | AK017844.1 | - | 3 - 4 | 775 | 82 | |
| AK089667.1 | - | 4 - 5 | 868 | 82 | ||
| AK156757.1 | - | 3 - 4 | 965 | 82 | ||
| BC100327.1 | - | 2 - 3 | 867 | 82 | ||
More Information
Set Membership: |
|

Ankrd33b
Genomic Map