Assay Details
| Gene Symbol: Cnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028044.2 | NP_082320.1 | 5 - 6 | 723 | 90 | |
| GenBank mRNA | AK005460.1 | - | 5 - 6 | 725 | 90 | |
| AK051121.1 | - | 4 - 5 | 941 | 90 | ||
| AK051628.1 | - | 5 - 6 | 741 | 90 | ||
| AK081672.1 | - | 5 - 6 | 638 | 90 | ||
| AK159555.1 | - | 5 - 6 | 768 | 90 | ||
| AK159871.1 | - | 5 - 6 | 753 | 90 | ||
| AK166578.1 | - | 5 - 6 | 731 | 90 | ||
| AK167201.1 | - | 5 - 6 | 744 | 90 | ||
| AK169021.1 | - | 5 - 6 | 748 | 90 | ||
| AK169098.1 | - | 5 - 6 | 768 | 90 | ||
| AK169410.1 | - | 5 - 6 | 698 | 90 | ||
| BC005788.1 | - | 1 - 2 | 50 | 90 | ||
| BC039643.1 | - | 1 - 2 | 118 | 90 | ||
| BC055711.1 | - | 5 - 6 | 724 | 6415164 | 90 | |
| BC085268.1 | - | 5 - 6 | 501 | 6487559 | 90 | |
More Information
Set Membership: |
|

Cnn3
Genomic Map