Assay Details
| Gene Symbol: Chst14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028117.3 | NP_082393.3 | 1 | 1223 | 64 | |
| GenBank mRNA | AK011230.1 | - | 1 | 1160 | 64 | |
| AK154555.1 | - | 1 | 1212 | 64 | ||
| AK154636.1 | - | 1 | 1192 | 64 | ||
| AK155115.1 | - | 2 | 1131 | 64 | ||
| AK159357.1 | - | 1 | 1191 | 64 | ||
| AK171614.1 | - | 1 | 1093 | 64 | ||
| AK208059.1 | - | 1 | 55 | 64 | ||
| BC026886.1 | - | 1 | 1044 | 64 | ||
| BC043700.1 | - | 1 | 1196 | 5346204 | 64 | |
| BC085479.1 | - | 1 | 1140 | 6415106 | 64 | |
More Information
Set Membership: |
|

Chst14
Genomic Map