Assay Details
| Gene Symbol: Cldn34c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028426.1 | NP_082702.1 | 2 - 3 | 201 | 98 | |
| GenBank mRNA | AK014019.1 | - | 2 - 3 | 169 | 98 | |
| BC027572.1 | - | 2 - 3 | 201 | 1363266 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014019.1, BC027572.1, NM_028426.1 transcripts.
Set Membership: |
|

Cldn34c1
Genomic Map