Assay Details
| Gene Symbol: Rap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028712.4 | NP_082988.1 | 1 | 4122 | 79 | |
| GenBank mRNA | AK151673.1 | - | 576 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rap2b| Gene Symbol: Rap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028712.4 | NP_082988.1 | 1 | 4122 | 79 | |
| GenBank mRNA | AK151673.1 | - | 576 | 79 | ||
Set Membership: |
|