Assay Details
| Gene Symbol: Efhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028889.2 | NP_083165.1 | 2 - 3 | 543 | 73 | |
| GenBank mRNA | AK016491.1 | - | 2 - 3 | 543 | 73 | |
| AK045911.1 | - | 2 - 3 | 545 | 73 | ||
| AK144175.1 | - | 2 - 3 | 546 | 73 | ||
| AK171846.1 | - | 2 - 3 | 545 | 73 | ||
| BC019531.1 | - | 2 - 3 | 529 | 4459208 | 73 | |
| BC085088.1 | - | 2 - 3 | 528 | 6467625 | 73 | |
More Information
Set Membership: |
|

Efhd1
Genomic Map