Assay Details
| Gene Symbol: Ptchd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029049.1 | NP_083325.1 | 3 - 4 | 1310 | 90 | |
| GenBank mRNA | AB235902.1 | - | 3 - 4 | 1225 | 90 | |
| AK015440.1 | - | 3 - 4 | 1269 | 90 | ||
| BC145091.1 | - | 3 - 4 | 1310 | 9053639 | 90 | |
| BC171943.1 | - | 3 - 4 | 1310 | 9054229 | 90 | |
More Information
Set Membership: |
|

Ptchd3
Genomic Map