Assay Details
| Gene Symbol: Rap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029519.3 | NP_083795.2 | 1 - 2 | 582 | 66 | |
| GenBank mRNA | AK075712.1 | - | 1 - 2 | 542 | 66 | |
| AK140318.1 | - | 1 - 2 | 594 | 66 | ||
| BC043066.1 | - | 1 - 2 | 553 | 5709337 | 66 | |
| BC049084.1 | - | 1 - 2 | 582 | 5706690 | 66 | |
| BC053003.1 | - | 1 - 2 | 582 | 5706690 | 66 | |
More Information
Set Membership: |
|

Rap2a
Genomic Map