Assay Details
| Gene Symbol: Ly6k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029627.2 | NP_083903.1 | 1 - 2 | 197 | 97 | |
| GenBank mRNA | AK010485.1 | - | 1 - 2 | 201 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010485.1, NM_029627.2 transcripts.
Set Membership: |
|

Ly6k
Genomic Map