Assay Details
| Gene Symbol: Ly6k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029627.2 | NP_083903.1 | 3 - 4 | 380 | 84 | |
| GenBank mRNA | AK010485.1 | - | 3 - 4 | 384 | 84 | |
| BC049723.1 | - | 2 - 3 | 219 | 6743692 | 84 | |
More Information
Set Membership: |
|

Ly6k
Genomic Map