Assay Details
| Gene Symbol: Tspyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029836.3 | NP_084112.1 | 4 - 5 | 1280 | 60 | |
| GenBank mRNA | AK018700.1 | - | 4 - 5 | 1222 | 60 | |
| AY273809.1 | - | 4 - 5 | 1121 | 60 | ||
| BC004006.1 | - | 4 - 5 | 928 | 60 | ||
| BC030334.1 | - | 2 - 3 | 427 | 60 | ||
| BC043694.1 | - | 3 - 4 | 1141 | 60 | ||
| BC054393.1 | - | 4 - 5 | 1200 | 6491672 | 60 | |
More Information
Set Membership: |
|

Tspyl2
Genomic Map