Assay Details
| Gene Symbol: Ddx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029936.2 | NP_084212.2 | 14 - 15 | 2203 | 158 | |
| GenBank mRNA | AK019495.1 | - | 14 - 15 | 2183 | 158 | |
| AK166787.1 | - | 2 - 3 | 402 | 158 | ||
| BC049261.1 | - | 14 - 15 | 2144 | 158 | ||
| BC080729.1 | - | 14 - 15 | 2171 | 6504816 | 158 | |
More Information
Set Membership: |
|

Ddx10
Genomic Map