Assay Details
| Gene Symbol: Ddx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029936.2 | NP_084212.2 | 2 - 3 | 352 | 91 | |
| GenBank mRNA | AK019495.1 | - | 2 - 3 | 332 | 91 | |
| AK045032.1 | - | 2 - 3 | 317 | 91 | ||
| AK052540.1 | - | 2 - 3 | 332 | 91 | ||
| BC023303.1 | - | 2 - 3 | 300 | 91 | ||
| BC049261.1 | - | 2 - 3 | 293 | 91 | ||
| BC055481.1 | - | 1 - 2 | 70 | 91 | ||
| BC080729.1 | - | 2 - 3 | 6504816 | 92 | ||
More Information
Set Membership: |
|

Ddx10
Genomic Map