Assay Details
| Gene Symbol: Ddx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029936.2 | NP_084212.2 | 6 - 7 | 958 | 86 | |
| GenBank mRNA | AK019495.1 | - | 6 - 7 | 938 | 86 | |
| AK045032.1 | - | 6 - 7 | 923 | 86 | ||
| AK052540.1 | - | 6 - 7 | 938 | 86 | ||
| BC023303.1 | - | 6 - 7 | 906 | 86 | ||
| BC049261.1 | - | 6 - 7 | 899 | 86 | ||
| BC055481.1 | - | 5 - 6 | 676 | 86 | ||
| BC080729.1 | - | 6 - 7 | 926 | 6504816 | 86 | |
More Information
Set Membership: |
|

Ddx10
Genomic Map