Assay Details
Gene Symbol: Lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033322.2 | NP_201579.1 | 7 - 8 | 624 | 98 | |
GenBank mRNA | AJ297352.1 | - | 7 - 8 | 620 | 98 | |
AK029671.1 | - | 7 - 8 | 656 | 98 | ||
AK030068.1 | - | 7 - 8 | 636 | 98 | ||
AK041091.1 | - | 7 - 8 | 657 | 98 | ||
AK046515.1 | - | 7 - 8 | 613 | 98 | ||
AK054003.1 | - | 7 - 8 | 630 | 98 | ||
AK081529.1 | - | 5 - 6 | 409 | 98 | ||
AK145263.1 | - | 7 - 8 | 633 | 98 | ||
AK147312.1 | - | 6 - 7 | 568 | 98 | ||
AK157770.1 | - | 7 - 8 | 631 | 98 | ||
BC026840.1 | - | 7 - 8 | 624 | 4165152 | 98 | |
BC037045.1 | - | 7 - 8 | 624 | 98 | ||
BC098224.1 | - | 7 - 8 | 629 | 1263280 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
