Assay Details
| Gene Symbol: Lmx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033652.5 | NP_387501.1 | 2 - 3 | 486 | 80 | |
| GenBank mRNA | AF226662.1 | - | 2 - 3 | 269 | 80 | |
| AK044944.1 | - | 2 - 3 | 488 | 80 | ||
| BC109167.2 | - | 2 - 3 | 407 | 40048180 | 80 | |
| BC109168.2 | - | 2 - 3 | 407 | 40048181 | 80 | |
More Information
Set Membership: |
|

Lmx1a
Genomic Map