Assay Details
| Gene Symbol: Bmp2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080708.1 | NP_542439.1 | 4 - 5 | 776 | 99 | |
| GenBank mRNA | AK046752.1 | - | 4 - 5 | 741 | 99 | |
| AY050249.1 | - | 4 - 5 | 776 | 99 | ||
More Information
Set Membership: |
|

Bmp2k
Genomic Map