Assay Details
| Gene Symbol: Anp32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130889.2 | NP_570959.1 | 6 - 7 | 931 | 105 | |
| GenBank mRNA | AB025582.1 | - | 6 - 7 | 752 | 105 | |
| AK165807.1 | - | 6 - 7 | 1001 | 105 | ||
| BC003489.1 | - | 6 - 7 | 949 | 2811622 | 105 | |
| BC005628.1 | - | 6 - 7 | 957 | 3963541 | 105 | |
| BC093506.1 | - | 6 - 7 | 963 | 4917194 | 105 | |
More Information
Set Membership: |
|

Anp32b
Genomic Map