Assay Details
| Gene Symbol: Slc25a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133668.3 | NP_598429.1 | 1 - 2 | 39 | 69 | |
| GenBank mRNA | AK028313.1 | - | 1 - 2 | 101 | 69 | |
| AK077274.1 | - | 1 - 2 | 40 | 69 | ||
| AK077723.1 | - | 1 - 2 | 96 | 69 | ||
| AK088013.1 | - | 1 - 2 | 44 | 69 | ||
| AK150641.1 | - | 1 - 2 | 44 | 69 | ||
| AK150729.1 | - | 1 - 2 | 44 | 69 | ||
| AK150870.1 | - | 1 - 2 | 83 | 69 | ||
| AK151088.1 | - | 1 - 2 | 44 | 69 | ||
| AK151518.1 | - | 1 - 2 | 44 | 69 | ||
| AK151887.1 | - | 1 - 2 | 44 | 69 | ||
| AK152006.1 | - | 1 - 2 | 44 | 69 | ||
| AK152292.1 | - | 1 - 2 | 44 | 69 | ||
| AK152341.1 | - | 1 - 2 | 40 | 69 | ||
| AK152713.1 | - | 1 - 2 | 44 | 69 | ||
| AK167197.1 | - | 1 - 2 | 44 | 69 | ||
| AK168129.1 | - | 1 - 2 | 85 | 69 | ||
| AK168580.1 | - | 1 - 2 | 40 | 69 | ||
| BC018161.1 | - | 1 - 2 | 54 | 2650436 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077274.1, AK077723.1, AK088013.1, AK150641.1, AK150729.1, AK150870.1, AK151088.1, AK151518.1, AK151887.1, AK152006.1, AK152292.1, AK152341.1, AK167197.1, AK168129.1, AK168580.1, BC018161.1, NM_133668.3 transcripts.
Set Membership: |
|

Slc25a3
Genomic Map