Assay Details
| Gene Symbol: Papd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133905.2 | NP_598666.1 | 6 - 7 | 891 | 88 | |
| GenBank mRNA | BC016629.1 | - | 6 - 7 | 891 | 3595512 | 88 |
More Information
Set Membership: |
|

Papd4
Genomic Map