Assay Details
| Gene Symbol: Otub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134150.2 | NP_598911.1 | 2 - 3 | 150 | 67 | |
| GenBank mRNA | AK027996.1 | - | 2 - 3 | 137 | 67 | |
| AK087195.1 | - | 2 - 3 | 137 | 67 | ||
| AK140070.1 | - | 2 - 3 | 137 | 67 | ||
| AK145273.1 | - | 2 - 3 | 140 | 67 | ||
| AK145970.1 | - | 2 - 3 | 142 | 67 | ||
| BC022575.1 | - | 2 - 3 | 150 | 5005025 | 67 | |
| BC054410.1 | - | 2 - 3 | 118 | 67 | ||
More Information
Set Membership: |
|

Otub1
Genomic Map