Assay Details
| Gene Symbol: Pim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138606.2 | NP_613072.1 | 1 - 2 | 257 | 97 | |
| GenBank mRNA | AK043570.1 | - | 1 - 2 | 302 | 97 | |
| BC027376.1 | - | 1 - 2 | 177 | 4954711 | 97 | |
| L41495.1 | - | 1 - 2 | 257 | 97 | ||
More Information
Set Membership: |
|

Pim2
Genomic Map