Assay Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144491.2 | NP_652762.2 | 4 - 5 | 425 | 77 | |
| GenBank mRNA | AK082812.1 | - | 4 - 5 | 430 | 77 | |
| AK128996.1 | - | 416 | 77 | |||
| AY078170.1 | - | 4 - 5 | 416 | 77 | ||
| BB651982.1 | - | 4 - 5 | 430 | 77 | ||
| BC044663.1 | - | 4 - 5 | 396 | 77 | ||
| BC080761.1 | - | 4 - 5 | 401 | 77 | ||
More Information
Set Membership: |
|

Dph1
Genomic Map