Assay Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145507.2 | NP_663482.2 | 1 - 2 | 260 | 132 | |
| NM_177445.5 | NP_803228.2 | 1 - 2 | 260 | 132 | ||
| GenBank mRNA | AK077533.1 | - | 1 - 2 | 183 | 132 | |
| AK077820.1 | - | 1 - 2 | 214 | 132 | ||
| AK144470.1 | - | 1 - 2 | 205 | 132 | ||
| AK146350.1 | - | 1 - 2 | 204 | 132 | ||
| AK151027.1 | - | 1 - 2 | 204 | 132 | ||
| AK151897.1 | - | 1 - 2 | 171 | 132 | ||
| AK152304.1 | - | 1 - 2 | 171 | 132 | ||
| AK152562.1 | - | 1 - 2 | 171 | 132 | ||
| AK153256.1 | - | 1 - 2 | 171 | 132 | ||
| AK153299.1 | - | 1 - 2 | 171 | 132 | ||
| AK166009.1 | - | 1 - 2 | 171 | 132 | ||
| AK169247.1 | - | 1 - 2 | 171 | 132 | ||
| AK169716.1 | - | 1 - 2 | 148 | 132 | ||
| BB576692.2 | - | 1 - 2 | 214 | 132 | ||
| BC008638.1 | - | 1 - 2 | 158 | 3586278 | 132 | |
| BY276340.1 | - | 1 - 2 | 261 | 132 | ||
| BY308833.1 | - | 1 - 2 | 258 | 132 | ||
More Information
Set Membership: |
|

Dars
Genomic Map