Assay Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145507.2 | NP_663482.2 | 3 - 4 | 420 | 130 | |
| GenBank mRNA | AK077820.1 | - | 3 - 4 | 374 | 130 | |
| BB576692.2 | - | 3 - 4 | 374 | 130 | ||
More Information
Set Membership: |
|

Dars
Genomic Map