Assay Details
| Gene Symbol: Ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145517.4 | NP_663492.3 | 3 - 4 | 426 | 107 | |
| GenBank mRNA | AK050385.1 | - | 3 - 4 | 411 | 107 | |
| BC012315.1 | - | 3 - 4 | 381 | 3981845 | 107 | |
| BC023695.1 | - | 3 - 4 | 406 | 5344622 | 107 | |
| BC023903.1 | - | 3 - 4 | 369 | 107 | ||
| BC025572.1 | - | 3 - 4 | 402 | 5323819 | 107 | |
| CN705165.1 | - | 3 - 4 | 426 | 107 | ||
More Information
Set Membership: |
|

Ormdl1
Genomic Map