Assay Details
| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145552.2 | NP_663527.2 | 3 - 4 | 363 | 73 | |
| GenBank mRNA | AK037796.1 | - | 3 - 4 | 299 | 73 | |
| AK077532.1 | - | 3 - 4 | 333 | 73 | ||
| AK146404.1 | - | 3 - 4 | 360 | 73 | ||
| BC003262.1 | - | 3 - 4 | 339 | 3501393 | 73 | |
| U69600.1 | - | 3 - 4 | 258 | 73 | ||
More Information
Set Membership: |
|

Gnl2
Genomic Map