Assay Details
| Gene Symbol: Dennd6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134465.2 | NP_001127937.1 | 7 - 8 | 777 | 67 | |
| NM_001285466.1 | NP_001272395.1 | 5 - 6 | 623 | 67 | ||
| NM_001285467.1 | NP_001272396.1 | 7 - 8 | 838 | 67 | ||
| NM_145969.4 | NP_666081.1 | 7 - 8 | 777 | 67 | ||
| GenBank mRNA | AK033036.1 | - | 5 - 6 | 625 | 67 | |
| AK054197.1 | - | 7 - 8 | 721 | 67 | ||
| AK054295.1 | - | 7 - 8 | 778 | 67 | ||
| AK131163.1 | - | 7 - 8 | 749 | 67 | ||
| AK132219.1 | - | 7 - 8 | 774 | 67 | ||
| AK135953.1 | - | 7 - 8 | 789 | 67 | ||
| AK161908.1 | - | 7 - 8 | 747 | 67 | ||
| AK163396.1 | - | 7 - 8 | 837 | 67 | ||
| BC025603.1 | - | 7 - 8 | 772 | 5250476 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033036.1, AK135953.1, AK163396.1, NM_001285466.1, NM_001285467.1 transcripts.
Set Membership: |
|

Dennd6a
Genomic Map