Assay Details
Gene Symbol: Hmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145998.3 | NP_666110.1 | 1 - 2 | 511 | 117 | |
GenBank mRNA | AK053014.1 | - | 1 - 2 | 281 | 117 | |
AK138228.1 | - | 1 - 2 | 512 | 117 | ||
BC023402.1 | - | 2 - 3 | 381 | 5043712 | 117 | |
S80989.1 | - | 1 - 2 | 305 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
