Assay Details
| Gene Symbol: Tmem185b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146103.2 | NP_666215.1 | 1 | 1318 | 89 | |
| GenBank mRNA | AK010846.1 | - | 1 | 1034 | 89 | |
| AK031130.1 | - | 1 | 1314 | 89 | ||
| BC020408.1 | - | 1 | 1306 | 89 | ||
| BC024712.1 | - | 1 | 1305 | 2646072 | 89 | |
| BC051523.1 | - | 1 | 175 | 89 | ||
More Information
Set Membership: |
|

Tmem185b
Genomic Map