Assay Details
| Gene Symbol: Pdik1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146156.3 | NP_666268.1 | 1 - 2 | 219 | 95 | |
| GenBank mRNA | AK164975.1 | - | 1 - 2 | 271 | 95 | |
| BC058514.1 | - | 1 - 2 | 205 | 6821637 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164975.1, BC058514.1, NM_146156.3 transcripts.
Set Membership: |
|

Pdik1l
Genomic Map