Assay Details
| Gene Symbol: Olfr693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146453.2 | NP_666664.1 | 1 | 723 | 95 | |
| GenBank mRNA | BC130248.1 | - | 1 | 726 | 40142368 | 95 |
More Information
Set Membership: |
|

Olfr693
Genomic Map