Assay Details
Gene Symbol: Olfr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146711.2 | NP_666922.2 | 1 | 1015 | 66 | |
GenBank mRNA | BC125464.1 | - | 1 | 40129979 | 66 | |
BC125468.1 | - | 1 | 40129983 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
