Assay Details
| Gene Symbol: Gjd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153086.5 | NP_694726.2 | 1 - 2 | 260 | 81 | |
| GenBank mRNA | AJ414562.1 | - | 1 - 2 | 69 | 81 | |
| AK034663.1 | - | 1 - 2 | 261 | 81 | ||
| AK053016.1 | - | 1 - 2 | 212 | 81 | ||
| BC137984.1 | - | 1 - 2 | 237 | 8861003 | 81 | |
More Information
Set Membership: |
|

Gjd4
Genomic Map