Assay Details
| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153591.4 | NP_705819.3 | 4 - 5 | 791 | 86 | |
| GenBank mRNA | AK046799.1 | - | 4 - 5 | 792 | 86 | |
| AK046801.1 | - | 4 - 5 | 792 | 86 | ||
| AK047091.1 | - | 4 - 5 | 736 | 86 | ||
| AK050718.1 | - | 4 - 5 | 730 | 86 | ||
| AK138260.1 | - | 4 - 5 | 771 | 86 | ||
| AK164179.1 | - | 4 - 5 | 760 | 86 | ||
More Information
Set Membership: |
|

Nars2
Genomic Map