Assay Details
| Gene Symbol: Zfp692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040686.1 | NP_001035776.1 | 10 - 11 | 1348 | 67 | |
| NM_182996.3 | NP_892041.2 | 10 - 11 | 1410 | 67 | ||
| GenBank mRNA | AK031177.1 | - | 9 - 10 | 1638 | 67 | |
| AK154896.1 | - | 1352 | 67 | |||
| AK170229.1 | - | 9 - 10 | 1701 | 67 | ||
| BC044879.1 | - | 4 - 5 | 1052 | 67 | ||
| BC055041.1 | - | 8 - 9 | 1747 | 67 | ||
| BC145470.1 | - | 10 - 11 | 1307 | 9054032 | 67 | |
| BC150776.1 | - | 10 - 11 | 1307 | 9087687 | 67 | |
More Information
Set Membership: |
|

Zfp692
Genomic Map