Assay Details
| Gene Symbol: Rbm15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045807.1 | NP_001039272.1 | 1 - 2 | 3074 | 80 | |
| GenBank mRNA | AK048987.1 | - | 2309 | 80 | ||
| AK077048.1 | - | 1 - 2 | 3464 | 80 | ||
| AK168242.1 | - | 1 - 2 | 2835 | 80 | ||
| AK168936.1 | - | 1 - 2 | 2959 | 80 | ||
| BC028452.1 | - | 1 - 2 | 632 | 80 | ||
| BC051409.1 | - | 1 - 2 | 2828 | 80 | ||
| BC057038.1 | - | 1 - 2 | 2838 | 80 | ||
| BC080828.1 | - | 1 - 2 | 2819 | 80 | ||
| BC120590.1 | - | 1 - 2 | 3074 | 40129513 | 80 | |
| BC137741.1 | - | 1 - 2 | 3074 | 8860760 | 80 | |
More Information
Set Membership: |
|

Rbm15
Genomic Map