Assay Details
| Gene Symbol: Ttc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080949.2 | NP_001074418.1 | 3 - 4 | 430 | 66 | |
| NM_177625.4 | NP_808293.1 | 3 - 4 | 430 | 66 | ||
| GenBank mRNA | AK044781.1 | - | 3 - 4 | 420 | 66 | |
| AK081214.1 | - | 3 - 4 | 406 | 66 | ||
| AK133953.1 | - | 3 - 4 | 412 | 66 | ||
| AK157777.1 | - | 3 - 4 | 409 | 66 | ||
| AK161279.1 | - | 3 - 4 | 408 | 66 | ||
| BC003272.1 | - | 3 - 4 | 400 | 66 | ||
| BC025610.1 | - | 3 - 4 | 412 | 5257921 | 66 | |
| BC069946.1 | - | 3 - 4 | 411 | 66 | ||
| BC092074.1 | - | 3 - 4 | 404 | 4920436 | 66 | |
More Information
Set Membership: |
|

Ttc5
Genomic Map