Assay Details
| Gene Symbol: Nlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081280.1 | NP_001074749.1 | 17 - 18 | 3476 | 73 | |
| NM_175547.3 | NP_780756.1 | 14 - 15 | 1399 | 73 | ||
| GenBank mRNA | AK084199.1 | - | 14 - 15 | 1313 | 73 | |
More Information
Set Membership: |
|

Nlrc3
Genomic Map