Assay Details
| Gene Symbol: Cystm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081365.1 | NP_001074834.1 | 1 - 2 | 186 | 68 | |
| GenBank mRNA | AK002512.1 | - | 60 | 68 | ||
| AK028217.1 | - | 57 | 68 | |||
| AK089711.1 | - | 1 - 2 | 48 | 68 | ||
| AK167679.1 | - | 91 | 68 | |||
| BC028765.1 | - | 113 | 1195860 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028217.1, AK167679.1, BC028765.1, NM_001081365.1 transcripts.
Set Membership: |
|

Cystm1
Genomic Map