Assay Details
| Gene Symbol: Galnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081421.1 | NP_001074890.1 | 6 - 7 | 874 | 108 | |
| GenBank mRNA | AB045325.1 | - | 6 - 7 | 836 | 108 | |
| AB175683.1 | - | 6 - 7 | 695 | 108 | ||
| AK138883.1 | - | 6 - 7 | 829 | 108 | ||
| AK173105.1 | - | 7 - 8 | 986 | 108 | ||
| BC110634.1 | - | 7 - 8 | 969 | 40059147 | 108 | |
| BC125015.1 | - | 7 - 8 | 968 | 40059141 | 108 | |
More Information
Set Membership: |
|

Galnt16
Genomic Map