Assay Details
| Gene Symbol: Aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172532.3 | NP_766120.1 | 6 - 7 | 1066 | 106 | |
| GenBank mRNA | AK052703.1 | - | 6 - 7 | 1067 | 106 | |
| AK144030.1 | - | 6 - 7 | 1050 | 106 | ||
| AK159485.1 | - | 6 - 7 | 1047 | 106 | ||
| AK166313.1 | - | 6 - 7 | 1046 | 106 | ||
| BC138703.1 | - | 6 - 7 | 1029 | 8861725 | 106 | |
More Information
Set Membership: |
|

Aldh5a1
Genomic Map