Assay Details
| Gene Symbol: Kcnh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172805.3 | NP_766393.2 | 3 - 4 | 1114 | 105 | |
| GenBank mRNA | AF309565.1 | - | 2 - 3 | 217 | 105 | |
| AK032438.1 | - | 3 - 4 | 1115 | 105 | ||
| BC066138.1 | - | 3 - 4 | 456 | 30094199 | 105 | |
More Information
Set Membership: |
|

Kcnh5
Genomic Map