Assay Details
| Gene Symbol: Kcnh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172805.3 | NP_766393.2 | 4 - 5 | 1237 | 101 | |
| GenBank mRNA | AF309565.1 | - | 3 - 4 | 340 | 101 | |
| AK032438.1 | - | 4 - 5 | 1238 | 101 | ||
| BC066138.1 | - | 4 - 5 | 579 | 30094199 | 101 | |
More Information
Set Membership: |
|

Kcnh5
Genomic Map