Assay Details
| Gene Symbol: Kcnh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172805.3 | NP_766393.2 | 8 - 9 | 2375 | 70 | |
| GenBank mRNA | AK032438.1 | - | 8 - 9 | 2376 | 70 | |
| BC066138.1 | - | 8 - 9 | 1717 | 30094199 | 70 | |
More Information
Set Membership: |
|

Kcnh5
Genomic Map