Assay Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173011.2 | NP_766599.2 | 1 - 2 | 191 | 108 | |
| GenBank mRNA | AF212319.1 | - | 1 - 2 | 162 | 108 | |
| AK088110.1 | - | 1 - 2 | 163 | 108 | ||
| AK145753.1 | - | 1 - 2 | 136 | 108 | ||
| AK146564.1 | - | 1 - 2 | 174 | 108 | ||
| AK161640.1 | - | 1 - 2 | 175 | 108 | ||
| AK169395.1 | - | 1 - 2 | 188 | 108 | ||
| BC060030.1 | - | 1 - 2 | 179 | 6515657 | 108 | |
| BY311397.1 | - | 1 - 2 | 200 | 108 | ||
More Information
Set Membership: |
|

Idh2
Genomic Map