Assay Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175200.4 | NP_780409.2 | 1 - 2 | 402 | 73 | |
| GenBank mRNA | AK076617.1 | - | 1 - 2 | 404 | 73 | |
| BB614746.1 | - | 1 - 2 | 403 | 73 | ||
| BC118929.2 | - | 1 - 2 | 368 | 40090362 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076617.1, BC118929.2 transcripts.
Set Membership: |
|

Als2cr11
Genomic Map